Learn More
Lamin A/C R482W Monoclonal Antibody (5H8-B4), Active Motif™
SDP
Mouse Monoclonal Antibody
Supplier:  Active Motif 39964
Encompass
 View more versions of this product
Catalog No. 50-199-3006
Add to cart
Description
Description
Lamins are a class of intermediate filament proteins that form a matrix on the inner surface of the nuclear envelope. These proteins are found in many different cell types in three different forms (A, B, and C). Lamins A and C are alternatively spliced versions of the LMNA gene. The LMNA gene has been linked to many disorders of the muscular system, nervous system, and the fat distributions systems including: Emery-Dreifuss muscular dystrophy, Dunnigan-type familial partial lipodystrophy (FPLD), limb-girdle muscular dystrophy (LGMD1B), dilated cardiomyopathy (CMD1A), axonal neuropathy (Charcot-Marie-Tooth disease; CMT2B1), and mandibuloacral dysplasia (MAD).
Specifications
Specifications
Lamin A/C R482W
Monoclonal
1 mg/mL
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
P02545
Mouse
Protein G
RUO
4000
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
5H8-B4
Unconjugated
LMNA
LMNA
This Lamin A/C R482W antibody was raised against a recombinant protein corresponding to amino acids 430-545 of human Lamin A/C containing the R482W substitution.
10 μg
Primary
Human
Antibody
IgG1