Learn More
Lamin A Mouse anti-Human, Mouse, Clone: 4A4-A4, eBioscience™ Invitrogen™
Mouse Monoclonal Antibody
Manufacturer: LIFE TECHNOLOGIES 14984882

Description
This 4A4-A4 monoclonal antibody recognizes human and mouse mature Lamin A. Applications Reported: This 4A4-A4 antibody has been reported for use in western blotting, immunocytochemistry and immunoprecipitation. Applications Tested: This 4A4-A4 antibody has been tested by western blotting of human cell line lysates. This may be used at 0.2 μg/mL. This 4A4-A4 antibody has also been tested by immunocytochemistry on A549 cells. This may be used at 0.06 μg/mL.
Lamin A Monoclonal antibody specifically detects Lamin A in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence, Western BlotSpecifications
Lamin A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P02545, P48678 | |
Mouse | |
Affinity chromatography | |
RUO | |
16905, 4000 | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence, Western Blot | |
4A4-A4 | |
Unconjugated | |
LMNA | |
70 kDa lamin; CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; lamin; lamin A; lamin A/C; lamin A/C-like 1; lamin C; lamin C2; lamin-A/C; LDP1; LFP; LGMD1B; LMN1; Lmna; LMNA protein; LMNC; LMNL1; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; Prelamin-A/C; PRO1; Renal carcinoma antigen NY-REN-32; unnamed protein product | |
KLH-conjugated peptide corresponding to the C-terminus of human Lamin A | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.