Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B1, Mouse anti-Human,Mouse, Biotin, Clone: 919022, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals MAB85251B

Catalog No. NB003248


Add to cart

Description

Description

Lamin B1 Monoclonal specifically detects Lamin B1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Lamin B1
919022
Western Blot, Immunohistochemistry
Purified, Conjugated
LMNB1
E. coli-derived partial recombinant human Lamin B1 protein (amino acids 471-582)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Western Blot, Immunohistochemistry
Biotin
PBS with 0.05% Sodium Azide
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419
Mouse
IgG2b
0.1 mL
Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Phospho Specific, Stem Cell Markers, Tumor Suppressors
Primary
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit