Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Clear Double-Sided Laminates

Double-sided/Industrial use

Manufacturer:  Brady™ 20589

 View more versions of this product

Catalog No. 19-104-507


Add to cart

Description

Description

  • Laminates documents 8.6 in. wide and up to 100 ft. long
  • Constructed from tough polyester
  • Available in sizes 8.6 in. x 50 ft. and 8.6 in. x 100 ft.
Specifications

Specifications

Safety
8.6 in. x 50 ft.
Double-Sided Laminate
Clear
21.8 cm x 15.24m
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit