Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B2 Mouse anti-Human, Mouse, Rat, Zebrafish, Clone: D5-C2, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP266924

Catalog No. NBP266924


Add to Cart

Description

Description

Lamin B2 Monoclonal specifically detects Lamin B2 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

LaminB2
Monoclonal
2 mg/mL
Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunohistochemistry-Paraffin
lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Mouse
Protein A purified
Primary
Human, Mouse, Rat, Zebrafish
Purified
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
D5-C2
Unconjugated
PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
LMNB2
Synthetic peptide within Human Lamin B2 aa 571-620/ 620. (SwissProt: Q03252 Human)
100 μL
84823
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit