Learn More
Molecular Dimensions LAMINEX GLASS COVER
SDP
Supplier:  Molecular Dimensions MD1152
Encompass
Protein Crystallography. LAMINEX GLASS COVER (Pack of 10) For Lipidic Cubic Phase, the Laminex system is built up from lower support plates of glass or UV transparent plastic with an adhesive spacer. On to this base, glass, UV transparent plates or films can be applied to create 96 chambers for crystal growth.
Catalog No. NC9863549