Learn More
Invitrogen™ Laminin-332 Recombinant Superclonal™ Antibody (5HCLC)
Rabbit Recombinant Superclonal Antibody
Supplier: Invitrogen™ 711306
Description
This antibody is predicted to react with Monkey, Sheep and Cat. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
Specifications
Laminin-332 | |
Recombinant Superclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q16787 | |
LAMA3 | |
Protein corresponding to Human LAMA3 (aa 2500-2700). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
5HCLC | |
Unconjugated | |
LAMA3 | |
[a]3B; BM600; BM600 150kD subunit; E170; Epiligrin; Epiligrin 170 kDa subunit; epiligrin alpha 3 subunit; epiligrin subunit alpha; kalinin 165kD subunit; kalinin subunit alpha; LAM3, alpha-3 subunit; Lama3; Lama3B; laminin 5 alpha 3; laminin 5, alpha-3 subunit; laminin A3; laminin subunit alpha 3; laminin subunit alpha-3; laminin, alpha 3; laminin, alpha 3 (nicein (150kD), kalinin (165kD), BM600 (150kD), epilegrin); Laminin-5; laminin-5 alpha 3 chain; laminin-5 subunit alpha; Laminin-6 subunit alpha; laminin-7 subunit alpha; LAMNA; LOCS; nicein 150kD subunit; nicein subunit alpha; nicein, 150kDa | |
Rabbit | |
Protein A | |
RUO | |
3909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.