Learn More
Alfa Aesar™ Laminin (925-933)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J64340EXD
 View more versions of this product
Catalog No. AAJ64340EXD
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
967.05
0.5mg
Solid
RUO
Laminin (925-933)
HPLC
Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor
Cys-Asp-Pro-Gly-Tyr-Ile-Gly-Ser-Arg
-30° to -10°C