Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 4 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI5G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271113JF549

Catalog No. N271113J549


Add to cart

Description

Description

Laminin alpha 4 Monoclonal antibody specifically detects Laminin alpha 4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Laminin alpha 4
OTI5G8
Immunohistochemistry
Affinity Purified, Conjugated
LAMA4
Recombinant protein fragment corresponding to amino acids 25-270 of human Laminin alpha 4 (NP_001098676) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp686D23145, LAMA3LAMA4&asterisk;-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Mouse
IgG1
0.1 mL
Primary
3910.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit