Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Laminin B2/γ 1 Ab-1, Rat Monoclonal Antibody

Recommended for Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded), Immunofluorescence and Immunoprecipitation (Native and denatured), Thermo Scientific™ Laminin B2/γ1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ RT795P1

 View more versions of this product

Catalog No. RT795P1


Add to cart

Description

Description

Recommended for Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded), Immunofluorescence and Immunoprecipitation (Native and denatured), Thermo Scientific™ Laminin B2/γ1 Ab-1, Rat Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Rat

Clone: A5

Isotype: IgG2a/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: Not determined

Immunogen: Murine EHS laminin preparation

Molecular Weight: ~210kDa

Positive Control: LS174T cells, Normal colon or colon CA

Cellular Localization: Basement membrane Specificity of Ab-1 was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Ab-1 stains uniformly all human and murine basement membranes.

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen and acid-alcohol-fixed, paraffin-embedded)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Laminin B2/gamma1 Ab-1
Monoclonal
200µg/mL
Purified
Murine EHS laminin preparation
500μL
Extracellular Matrix
Primary
Human
Immunohistochemistry (Frozen), Immunoprecipitation
A5
Unconjugated
Rat
IgG2a kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit