Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Laminin B2/γ 1 Ab-2, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ MS1356R1

 View more versions of this product

Catalog No. MS1356R1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Recommended for Immunohistochemistry (Formalin/paraffin), Western Blotting (Not verified) and Immunoprecipitation (Not verified), Thermo Scientific™ Laminin B2/γ1 Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of α, β, and γ chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Host Species: Mouse

Clone: D18

Isotype: IgG2a/κ

Species Reactivity: Human, Rat and Guinea Pig. Others not known.

Epitope: Not determined

Immunogen: Rat glomerular basement membrane

Molecular Weight: ~210kDa

Positive Control: LS174T cells, Skin

Cellular Localization: Basement membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunoprecipitation (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Laminin B2/gamma1 Ab-2
Monoclonal
Unconjugated
Prediluted
Rat glomerular basement membrane
LS174T cells, Skin
RUO
Primary
Monoclonal
Immunohistochemistry (Paraffin)
D18
Ready-to-use for IHC (UVLP)
Murine
IgG2a, κ
1mL
Cancer and Tumor Biology
Primary
Guinea Pig, Human, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit