Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin beta-3 Mouse anti-Human, Clone: CL3363, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524655

Catalog No. PIMA524655


Add to cart

Description

Description

Immunogen sequence: KLVTSMTKQL GDFWTRMEEL RHQARQQGAE AVQAQQLAEG ASEQALSAQE GFERIKQKYA ELKDRLGQSS MLGEQGARIQ SVKTEAEELF GETMEMMDRM KDMELELLRG SQAIMLRSAD LTGLEKRVEQ Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%. Binds to an epitope located within the peptide sequence MTKQLGDFWTRMEEL as determined by overlapping synthetic peptides.

The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.
Specifications

Specifications

Laminin beta-3
Monoclonal
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q13751
RP1-272L16.3, BM600-125KDA, LAM5, LAMNB1, epiligrin subunit bata, kalinin B1 chain, kalinin subunit beta, kalinin-140kDa, laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), laminin-5 subunit beta, nicein subunit beta, nicein-125kDa
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3914
Immunohistochemistry, Western Blot
CL3363
Unconjugated
LAMB3
Liquid
LAMB3
Recombit protein corresponding to Human Laminin beta-3
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only