Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin Mouse anti-Human, Clone: 4C7, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$212.70 - $746.00
Specifications
Antigen | Laminin |
---|---|
Clone | 4C7 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB12017107
|
Novus Biologicals
NB12017107 |
0.5 mL |
Each for $746.00
|
|
NB018081
|
Novus Biologicals
NB120171070.1ML |
0.1 mL |
Each for $212.70
|
|
Description
Laminin Monoclonal specifically detects Laminin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Laminin | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
284217 | |
IgG2a κ | |
Store at 4°C. |
4C7 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
LAMA1 | |
Primary |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title