Learn More
Selleck Chemical LLC
Lamivudine 25mg 134678-17-4 GR109714X
Manufacturer: Selleck Chemical LLC S170625MG
Lamivudine (GR109714X) is a potent nucleoside analog reverse transcriptase inhibitor, used for treatment of chronic HBV and HIV/AIDS. It works by blocking the HIV reverse transcriptase and hepatitis B virus polymerase.