Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamotrigine isethionate, Tocris Bioscience™

Catalog Number p-4784096
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
113170-86-8
C11H13Cl2N5O4S
382.216
CJIDZLNMKONKAD-UHFFFAOYSA-N
lamotrigine isethionate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine isethionate, 3,5-diamino-6-2,3-dichlorophenyl-1,2,4-triazine isethionate, acmc-1bsdt, isethionic acid; lamotrigine, lamotrigine isethionate hplc, 2-hydroxyethane-1-sulfonic acid-6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 1/1, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 2-hydroxyethanesulfonate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine mono 2-hydroxyethanesulfonate
45039646
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine;2-hydroxyethanesulfonic acid
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N.C(CS(=O)(=O)O)O
This item is not returnable. View return policy
113170-86-8
C11H13Cl2N5O4S
382.216
CJIDZLNMKONKAD-UHFFFAOYSA-N
lamotrigine isethionate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine isethionate, 3,5-diamino-6-2,3-dichlorophenyl-1,2,4-triazine isethionate, acmc-1bsdt, isethionic acid; lamotrigine, lamotrigine isethionate hplc, 2-hydroxyethane-1-sulfonic acid-6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 1/1, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine 2-hydroxyethanesulfonate, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine mono 2-hydroxyethanesulfonate
45039646
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine;2-hydroxyethanesulfonic acid
C1=CC(=C(C(=C1)Cl)Cl)C2=C(N=C(N=N2)N)N.C(CS(=O)(=O)O)O

Inhibits glutamate release. Water-soluble salt of lamotrigine (Cat. No. 1611)

Water-soluble salt of lamotrigine (Cat. No. 1611). Displays anticonvulsant effects and inhibits glutamate release, possibly through inhibition of Na+, K+ and Ca2+ currents.
Quantity 10 mg
Formula Weight 382.22
Percent Purity >99%
Chemical Name or Material Lamotrigine isethionate
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit