LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage

Contain microbe-specific media enclosed in a sterile vial

Manufacturer: LaMotte™ 5541

 View more versions of this product

Catalog No. 03-420-893

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Sterile Yes
    Storage Requirements 18-25°C
    Color Yellow
    For Use With (Application) Food and beverage: isolation and differentiation of gram (-) enteric bacilli, Coliform testing/recovery of stressed Coliforms
    Includes Free basic microbe identification application and access to technical information sheet
    Manufacturing Origin United States
    Media Type NUT-TTC/EMB dual Agar
    Packaging Paddle
    Quantity 40mL
    Shelf Life 1 Year

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit