Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage Available on GSA/VA Contract for Federal Government customers only.
Contain microbe-specific media enclosed in a sterile vial
Manufacturer:  LaMotte™ 5552/RMD
 View more versions of this product
Catalog No. 03-420-899
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Offwhite
Free basic microbe identification application and access to technical information sheet
TSA/RB dual Agar
40mL
18-25°C
Food and beverage: cultivation of a wide variety of aerobic and anaerobic microorganisms (TSA), selective enumeration and cultivation of Yeasts, molds and Actinomycetes from food and other surfaces (RB)
United States
Paddle
Yes
Safety and Handling
ShelfLife : 1 Year