LaMotte™ BioPaddles™ Dual-Agar Paddles for Food and Beverage

Contain microbe-specific media enclosed in a sterile vial

Manufacturer: LaMotte™ 5540

 View all options for this product

Catalog No. 03-420-892


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color Pink
    For Use With (Application) Food and beverage: enumeration and cultivation of bacteria from potable Water, total Coliform testing (TCC), Coliform testing/recovery of stressed Coliforms
    Includes Free basic microbe identification application and access to technical information sheet
    Manufacturing Origin United States
    Media Type R2A/MAC dual Agar
    Packaging Paddle
    Quantity 40mL
    Shelf Life 1 Year
    Sterile Yes
    Storage Requirements 18-25°C

    Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.

    • Longer shelf life than traditional Petri dishes
    • No refrigeration required
    • No need for inoculating loops or Bunsen burners
    • Requires simple incubation and a magnifier
    • Unique design enhances colony morphology characterization
    • Ten paddles per box
    R2A/MacConkey: For the cultivation and enumeration of bacteria from potable water, total coliform testing (TCC).
    Nutrient-TTC/EMB: For the cultivation and enumeration of Coliform bacteria including TCC (Total Colony Counts). Non-selective, supports growth of a wide variety of organisms.
    LIS/EMB Agar: For enumeration and selective isolation of Listeria spp. and total coliform (TCC)
    HEK/SS: For enumeration and selective isolation of pathogenic intestinal bacteria.
    MacConkey/EMB:For isolation and differentiation of Gram (-) cocci enteric bacilli, coliform, and recovery of stressed coliforms.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit