Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Chlorine Test Kit

Measures free, total and combined chlorine levels

Manufacturer:  LaMotte™ 330801/DEL

Catalog No. 22-278-557

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Includes:
  • 50 Chlorine DPD #1R tablets
  • 50 Chlorine DPD #3R Tablets
  • Two test tubes, plastic, w/caps
  • One Octa-Slide 2 viewer
  • One Chlorine Octa-Slide 2 Bar, 0.2 to 3.0 ppm

Description

Description

  • Test Method: DPD Tablet and Octa-Slide 2 Comparator
  • Test Range: 0.2 to 3.0 ppm Cl
  • Number of Tests: 50
  • Number of Reagents per test: two

DPD = N,N Diethyl-1,4 Phenylenediamine Sulfate
PPM = Parts Per Million
Cl = chemical symbol for Chlorine

For testing chemical chlorine levels in drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater

Compliance

Accepted by the Environmental Protection Agency (EPA) for National Primary Drinking Water Regulation (NPDWR) monitoring.

Specifications

Specifications

Chemical Chlorine Test Kit
Drinking water, environmental education, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
50 Tests
NPDWR
50 chlorine DPD 1R tablets, 50 chlorine DPD 3R tablets, 2.5 to 5 to 10mL test tubes, with caps, Octa-slide 2 viewer, chlorine octa-slide 2 bar
DPD tablets and Octa-Slide 2 comparator
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For compliance monitoring a Check Standard should be prepared.