LaMotte™ SMART™ Spectro System COD Reagents Available on GSA/VA Contract for Federal Government customers only.

A precise and easy way to measure critical COD levels

Manufacturer: LaMotte™ 0075SC

 View more versions of this product

Catalog No. 22-307-877

  Check Availability
Add to Cart

Alert

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.


    Specifications

    Length (Metric) 1.6cm
    Test Method Acid digestion
    Certifications/Compliance EPA
    Range 0 to 150ppm
    For Use With (Equipment) LaMotte™ colorimeters
    Length (English) 0.62 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit