Learn More
LaMotte™ SMART™ Spectro System COD Reagents
A precise and easy way to measure critical COD levels
Manufacturer:  LaMotte™ 0076SC/DEL
 View more versions of this product
Catalog No. 22-307-878
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
Description
Measure low, medium or high levels of Chemical Oxygen Demand (COD) using a mercury reagent system approved by the Environmental Protection Agency (EPA).
  • Comes in 25 vials to a case
  • Simply add the test sample to the reagent in the vial and heat the vial for up to two hours at 150°C (±2°C)
  • Once the digestion is complete, place the tube directly into the meter and read the ppm COD
  • 0075-SC tests 0 to 150 ppm
  • 0076-SC tests 0 to 1500 ppm
  • 0077-SC tests 0 to 15,000 ppm
Use with Lamotte COD PLUS Colorimeter, SMART 2 Colorimeter™ or SMART Spectro Spectrophotometer.
Order Info
Each package contains 25 vials with PTFE-lined caps to offer the maximum level of safety during use. High grade, optical quality, standard size (16mm) vials are protected in a premium EPS foam shipper for full compliance to global shipping requirements. All kits ship as R1–Small Qty. Hazardous Material- No Fee.
Specifications
Specifications
EPA
LaMotte™ colorimeters
1.6cm
0 to 1500ppm
0.62 in.
Acid digestion