Learn More
LaMotte™ Chlorine Bleach Test Kit
Direct Reading Titrator - Test uses a 1.0mL microburet calibrated to allow direct reading of the test result within a specific range
Manufacturer:  LaMotte™ 710503/DEL
Catalog No. 22-307-845
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Chlorine Bleach, Iodometric Titration - Test uses an iodine titration to cover the range 0 to 10% chlorine bleach
  • 50 tests at 10% per kit
Specifications
Specifications
0 to 10% or 0.2%
Chlorine Bleach Kit
2 x 30mL hypochlorite reagent A, 10g potassium iodide crystals, 60mL hypochlorite reagent D, test tube with cap, direct reading titrator, 0.5mL plastic pipet, 1mL plastic pipet, plastic spoon
Measures for chlorine bleach in industrial water boiling and cooling, water and wastewater
Iodometric Titration
50.0