LaMotte™ COD Colorimeter Multi-Range Reagent Systems Available on GSA/VA Contract for Federal Government customers only.
Measure levels of Chemical Oxygen Demand using the SMART Spectro, the original SMART colorimeter or the SMART 2 colorimeter
Manufacturer: LaMotte™ 0073SC
 View more versions of this product
Catalog No. 22-307-881
  Check Availability
Add to Cart
    Specifications
    Range 0-1500ppm
    Includes 25 high grade, optical quality 16mm vials with TFE lined caps
    For Use With (Equipment) SMART spectro, original SMART or SMART2 colorimeters
    For Use With (Application) Measure levels of chemical oxygen demand
    Product Type Water Testing System
  • Description & Specifications
Related Products
Product Content Feedback