Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ DPD Free Chlorine Reagent Refills

The liquid alternative to DPD tablet refills

Manufacturer:  LaMotte™ P6740H/DEL

 View more versions of this product

Catalog No. 22-278-644

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »

Alert:

Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.


Description

Description

  • DPD £1A and DPD £1B test Free Available Chlorine (Cl)
  • DPD £3 is added to the reacted sample to measure Total Cl
  • Comes in 30mL and 60mL sizes

DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate

Use with existing LaMotte chlorine comparators or colorimeters. For each test, all three reagents are added to the same sample. Ideal for testing drinking water, food and beverage, pool and spa, industrial water boiling and cooling, laundry and sanitation, water and wastewater.

Specifications

Specifications

DPD 1A Liquid Reagent
LaMotte™ colorimeters and comparators
2 oz.
Measurement of total chlorine, provides quicker results and clearer colors than tablets
Each
60mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit