Learn More
LaMotte™ Dual Range Free and Total Chlorine Test Kit
Octa-Slide - Colorimetric tests use eight color standards on a comparator block for one-by-one viewing against the reacted sample
Manufacturer:  LaMotte™ 331401/DEL
Catalog No. 22-307-854
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Chlorine, Free and Total Kit, Dual Range - DPD method for Free Available Chlorine, Total Residual Chlorine and Combined Chlorine. Low range: 0.1 to 1.0ppm Cl; High range: 1.0 to 6.0ppm Cl. With two Chlorine Octa-Slide Bar (low range, high range). 100 tests per kit
  • Octa-slide color comparator included
Specifications
Specifications
Low range 0.1 to 10, high range 1 to 6ppm
Dual Range Free and Total Chlorine Kit
100.0
Measures for high and low ranges of chlorine in drinking water, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
DPD Reaction