Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Dual Range Phosphate Test Kit

Test system: Phosphomolybdate complex/Stannous Chloride reduction

Manufacturer:  LaMotte™ 440801/DEL

Catalog No. 22-307-851

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Simple colorimeter

  • Phosphate - Phosphomolybdate complex is reduced by ascorbic acid to produce a blue color
  • Range: 0 to 3.0ppm
  • 100 tests

Specifications

Specifications

Low range1 to 10, high range 10 to 100ppm
Dual Range Phosphate Test Kit
2 x 30mL VM phosphate reagent, 30mL reducing reagent, plastic pipet dropping, 1mL plastic pipet, 0.5mL plastic pipet, 2 test tubes with caps, phosphate octa-slide 2 Bar, octa-slide 2 viewer
Measures for industrial water boiling and cooling, laundry, sanitation, water and wastewater
Phosphomolybdate complex/Stannous Chloride reduction
50.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit