Learn More
LaMotte™ Green Advanced Water Monitoring Kit
Excellent environmental education tool for monitoring water quality
Manufacturer:  LaMotte™ 588401/DEL
Catalog No. 22-307-865
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: Contains all the LaMotte™ water quality test kits featured and supported in The Field Manual for Water Quality Monitoring (12th edition) by Mark Mitchell and William Stapp, a standard text for school-based water quality monitoring programs in schools around the world.
Description
Description
Teach students how to determine the properties of water utilizing a range of chemical- and instrument-based test factors.
Test factors in kit include:
  • Dissolved oxygen
  • Precision pH
  • Phosphate
  • Auxiliary phosphate
  • Nitrate-Nitrogen
  • Turbidity
  • Thermometer
For Grades 7 and up
Specifications
Specifications
Environmental education
Advanced Green Water Quality Monitoring Kit
Dissolved oxygen, precision pH, phosphate, auxiliary phosphate, nitrate nitrogen, turbidity thermometer, comes with field manual for water quality monitoring