LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips Available on GSA/VA Contract for Federal Government customers only.

For water quality testing

Manufacturer: LaMotte™ 3000

 View more versions of this product

Catalog No. 22-307-874

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Water quality testing
    Parameter Specifications Peracetic acid
    Quantity 50 Strips per Vial
    Range 0-10-20-40-60-85-160ppm
    Storage Requirements Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
    Product Type Single Factor Test Strips

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit