LaMotte™ Reagent Refills Available on GSA/VA Contract for Federal Government customers only.

Testing supplies for the discerning scientist

Manufacturer: LaMotte™ 6410L

 View more versions of this product

Catalog No. 03-920-465

  Check Availability
Add to Cart

    Specifications

    Includes Mixed acid nitrogen reducing reagent, alkalinity reagent B, chlorine reagent 1
    Storage Requirements 25°C
    Volume (English) 16.9 oz.
    Volume (Metric) 500mL
    Shelf Life 2 Years
    Product Type Ferroin Indicator

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit