LaMotte™ Reagent Refills Available on GSA/VA Contract for Federal Government customers only.

Testing supplies for the discerning scientist

Manufacturer: LaMotte™ 4493DRH

 View more versions of this product

Catalog No. 03-920-350

  Check Availability
Add to Cart

    Specifications

    pH Range 2
    Storage Requirements 25°C
    Volume (English) 2.02 oz.
    Volume (Metric) 60mL
    Shelf Life 2 Years
    Color Clear
    Composition 0.1% Sulfuric Acid, 100% Water
    Format Liquid
    Product Type Alkalinity Reagent B

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit