Learn More
LaMotte™ Series 1200 Single-Parameter Colorimeters
Water-resistant, handheld single-parameter meters provide fast, accurate, direct results in actual ppm concentrations with photometric accuracy to ±0.001 absorbance units
Specifications
Certifications/Compliance CE
View More Specs
Includes: Come in a sturdy carrying case with six screw cap sample vials, RS-232 port to interface with a datalogger or computer and an instruction manual.
Products 1
Catalog Number Mfr. No. Analytes Range Method No. of Tests Price Quantity & Availability  
Catalog Number Mfr. No. Analytes Range Method No. of Tests Price Quantity & Availability  
22-278-565
View Documents
LaMotte™
368001/DEL
Ammonia Nitrogen 0 to 5ppm Nessler Colorimeter 100.0 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Test for a variety of analytes using colorimetric single test kits or visual colorimetric/titration test kits.
  • Microprocessor allows preprogrammed calibrations to optimally match non-linear curves Autozero function
  • Flip-top lid over sample chamber eliminates problems with stray light and misplaced light caps
  • Chlorine colorimeter kits use unit-dose DPD tablets or DPD liquid reagents; all analytes have enough reagent for 100 tests
Order Info
Dimensions: 6L x 3.25W x 2.50 in. H (15 x 8 x 5.5cm). 110V 60Hz. All kits require a 9-volt DC battery. Optional AC adapter sold separately (contact your Fisher Scientific Customer Service Representative for details).
Compliance
CE Mark of Compliance for electromagnetic compatibility and safety.
† Refills available online; see 22-278-643, -645, and 647.
Specifications
Specifications
CE