Learn More
LaMotte™ Series 1200 Single-Parameter Colorimeters
Water-resistant, handheld single-parameter meters provide fast, accurate, direct results in actual ppm concentrations with photometric accuracy to ±0.001 absorbance units
Supplier: LaMotte™ 368001/DEL
Includes: Come in a sturdy carrying case with six screw cap sample vials, RS-232 port to interface with a datalogger or computer and an instruction manual.
Description
Test for a variety of analytes using colorimetric single test kits or visual colorimetric/titration test kits.
- Microprocessor allows preprogrammed calibrations to optimally match non-linear curves Autozero function
- Flip-top lid over sample chamber eliminates problems with stray light and misplaced light caps
- Chlorine colorimeter kits use unit-dose DPD tablets or DPD liquid reagents; all analytes have enough reagent for 100 tests
Order Info
Dimensions: 6L x 3.25W x 2.50 in. H (15 x 8 x 5.5cm). 110V 60Hz. All kits require a 9-volt DC battery. Optional AC adapter sold separately (contact your Fisher Scientific Customer Service Representative for details).
Compliance
CE Mark of Compliance for electromagnetic compatibility and safety.
Specifications
Ammonia Nitrogen | |
Industrial water boiling and cooling, water and wastewater | |
100 | |
Nessler Colorimeter |
CE | |
30mL ammonia nitrogen reagent, 3 x 30mL ammonia nitrogen reagent, 1 1mL pipet, plastic, 1 colorimeter tubes, with caps, water sample collecting bottle, 1200 colorimeter for ammonia nitrogen | |
0 to 5ppm | |
Ammonia Nitrogen Test Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.