LaMotte™ Series 1200 Single-Parameter Colorimeters Available on GSA/VA Contract for Federal Government customers only.
Water-resistant, handheld single-parameter meters provide fast, accurate, direct results in actual ppm concentrations with photometric accuracy to ±0.001 absorbance units
Manufacturer: LaMotte™ 367001
 View more versions of this product
Catalog No. 22-278-567
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Drinking water supplies and distribution systems, swimming pool and spas, sewage and chlorinated waste water, process water and sanitizing solutions
    Includes 100 chlorine DPD instrument grade tablets, colorimeter tubes, with caps, set of 6, 2 tablets crushers, water sample collecting bottle, 1200 colorimeter for chlorine DPD
    No. of Tests 100.0
    Method DPD Tablets Colorimeter
    Analytes Free and Total Chlorine
    Certifications/Compliance CE
    Product Type Chlorine Free and Total Test Kit
    Range 0 to 4ppm
  • Description & Specifications
Related Products
Product Content Feedback