Learn More
LaMotte™ Series 1200 Single-Parameter Colorimeters
Water-resistant, handheld single-parameter meters provide fast, accurate, direct results in actual ppm concentrations with photometric accuracy to ±0.001 absorbance units
Manufacturer:  LaMotte™ 367001LI
 View more versions of this product
Catalog No. 22-278-568
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Free and Total Chlorine
Chlorine Free and Total Test Kit
Drinking water supplies and distribution systems, swimming pool and spas, sewage and chlorinated waste water, process water and sanitizing solutions
140.0
CE
0 to 4ppm
30mL DPD free chlorine reagent, 30mL DPD 1B free chlorine reagent, 30mL DPD total chlorine reagent, colorimeter tubes, with caps, set of 6, water sample collecting bottle, 1200 colorimeter for chlorine DPD
DPD Liquid Colorimeter