LaMotte™ Series 1200 Single-Parameter Colorimeters Available on GSA/VA Contract for Federal Government customers only.
Water-resistant, handheld single-parameter meters provide fast, accurate, direct results in actual ppm concentrations with photometric accuracy to ±0.001 absorbance units
Manufacturer: LaMotte™ 367901
 View more versions of this product
Catalog No. 22-278-577
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Drinking, surface and saline water, domestic and industrial wastes (method based on reactions that are specific for orthophosphate)
    Includes 2 x 60mL phosphate acid reagent, 10g phosphate reducing reagent, 6 colorimeter tubes, 10mL, with cap, 1 0.1g spoon, plastic, water sample collecting bottle, 1mL pipet, plastic, 1200 colorimeter for phosphate
    No. of Tests 100.0
    Method Ascorbic Acid
    Analytes Phosphate
    Certifications/Compliance CE
    Product Type Phosphate Test Kit
    Range 0 to 3ppm
  • Description & Specifications
Related Products
Product Content Feedback