LaMotte™ SMART 3™ Colorimeter Reagent System Available on GSA/VA Contract for Federal Government customers only.
For water analysis in the field or in the lab
Manufacturer: LaMotte™ 3640SC
 View more versions of this product
Catalog No. 22-278-511
  Check Availability
Add to Cart
    Specifications
    Range 0-3.50ppm
    Test Method Bicinchoninic acid, 1 reagent
    For Use With (Equipment) SMART3 colorimeter
    For Use With (Application) Water analysis in the field or in the lab
    Product Type Water Testing Reagent
  • Description & Specifications
Related Products
Product Content Feedback