Learn More
LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems Products for Science Education
For water analysis in the field or in the lab
Manufacturer:  LaMotte™ 3650SC/DEL
 View more versions of this product
Catalog No. 22-278-523
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • High-quality reagents manufactured from ACS raw materials for high reproducibility
  • All systems are designed to overcome test interferences for fast and dependable results
Order Info
Additional reagent systems are available. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Specifications
Nitrate Nitrogen
Test Kit
0 to 0.8ppm
SMART3 colorimeter
Diazotization/coupling (2)
Nitrite Nitrogen
Water testing and environmental studies
20
Diazotization