Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems

For water analysis in the field or in the lab

Manufacturer:  LaMotte™ 366701SC/DEL

 View more versions of this product

Catalog No. 22-278-536

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Choose from the following test factors:

  • Zinc Laboratory Reagent (LR)
  • Manganese
  • Aluminum
  • Ozone
  • Cyanide
  • Phosphate
  • Dissolved Oxygent
  • Chromium Hexavalent
  • Ammonia Nitrogen LR

Order Info

Additional reagent systems are available. Contact your Fisher Scientific Customer Service Representative for details.

Specifications

Specifications

SMART™ spectro spectrophotometer
50
SMART Spectro Spectrophometer and SMART 3 Colorimeter
Zinc LR
6
Zinc, Low Range, LaMotte test reagent
0 to 3ppm, 0 to 3 ppm
Water testing and environmental studies
50
Zincon
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit