Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte SMART Spectro Spectrophometer and SMART 3 Colorimeter Reagent Systems
For water analysis in the field or in the lab
Specifications
For Use With (Application) | Water testing and environmental studies |
---|---|
For Use With (Equipment) | SMART3 colorimeter |
Catalog Number | Mfr. No. | Type | Range | Test Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Range | Test Method | Price | Quantity & Availability | |||
22-278-530
|
LaMotte
3639SC/DEL |
Potassium | 0 to 10ppm | Tetraphenylboron |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
22-278-528
![]() |
LaMotte
3655SC/DEL |
Phosphate | 0 to 70ppm | Molybdovanadate |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- High-quality reagents manufactured from ACS raw materials for high reproducibility
- All systems are designed to overcome test interferences for fast and dependable results
Order Info
Additional reagent systems are available. Contact your Fisher Scientific Customer Service Representative for details.
Specifications
Water testing and environmental studies | |
SMART3 colorimeter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title