LaMotte™ SMART 3™ Colorimeter Reagents Available on GSA/VA Contract for Federal Government customers only.
The perfect complement for your SMART 3 Colorimeter
Manufacturer: LaMotte™ 366001SC
 View more versions of this product
Catalog No. 22-278-512
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) SMART3 colorimeter
    No. of Tests 50.0
    Method Pyridine-Barbituric Acid (5)
    Analytes Cyanide
    Product Type Test Kit
    Range 0 to 0.5ppm
  • Description & Specifications
Related Products
Product Content Feedback