LaMotte™ SMART 3™ Colorimeter Reagents
The perfect complement for your SMART 3 Colorimeter
Manufacturer: LaMotte™ 3649-SC/DISC
 View more versions of this product
Catalog No. 22-278-522
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) SMART3 colorimeter
    No. of Tests 20.0
    Method Cadmium Reduction (2)
    Analytes Nitrate Nitrogen
    Product Type Test Kit
    Range 0 - 3.0ppm
  • Description & Specifications
Related Products
Product Content Feedback