LaMotte™ SMART 3™ Colorimeter Reagents Available on GSA/VA Contract for Federal Government customers only.
The perfect complement for your SMART 3 Colorimeter
Manufacturer: LaMotte™ 3655SC
 View more versions of this product
Catalog No. 22-278-528
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Equipment) SMART3 colorimeter
    No. of Tests 50.0
    Method Molybdovanadate (1)
    Analytes Phosphate
    Product Type Test Kit
    Range 0 to 70ppm
  • Description & Specifications
Related Products
Product Content Feedback