Learn More
LaMotte™ SMART 3™ Colorimeter Reagents Available on GSA/VA Contract for Federal Government customers only.
The perfect complement for your SMART 3 Colorimeter
Manufacturer:  LaMotte™ 3643SC
 View more versions of this product
Catalog No. 22-278-501
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Bromine/Chlorine/Iodine
Bromine 0 to 9, chlorine 0 to 4, iodine 0 to 14ppm
100.0
Test Kit
SMART3 colorimeter
DPD Tablets (2)