Learn More
LaMotte™ SMART™ Spectro Reagent Systems Products for Science Education
A wide range of reagent systems to meet your quality testing needs
Manufacturer:  LaMotte™ 3651SC
 View more versions of this product
Catalog No. 22-278-525
  Check Availability
Add to cart
Requires : Use with the Lamotte SMART 3 Colorimeter and the SMART Spectro Spectrophotometer.
Specifications Description & Specifications
Specifications
SMART™ spectro spectrophotometer
LaMotte test reagent
0 to 0.4ppm
Indigo Trisulfonate
100
Ozone LR
3