Learn More
LaMotte™ SMART™ Water Analysis Laboratory
SDPSCI_ED
The SMART™ Water Analysis Laboratory features a digital colorimeter that analyzes test sample color reactions and provides direct readouts for 15 factors.
Supplier:  LaMotte™ 195103
 View more versions of this product
Catalog No. S94987
Includes: Includes digital meters to measure pH, conductivity, and TDS
Description
Description
  • Portable.
  • Measures 24 water quality parameters for pollution detection, environmental studies, and industrial water and wastes.
  • Provide results directly in ppm for 6 additional factors.
  • Programmed to provide direct readouts for over 40 factors using optional reagent packages.
Specifications
Specifications
0 to 14
Pollution detection, environmental studies and industrial water and wastes
Digital meters to measure pH, conductivity, and TDS
SCL-05, 120 V/60 Hz
500 mL
SMART colorimeter
Water Analysis Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.