Learn More
LaMotte™ Total Dissolved and Hydrogen Sulfide Test Kit
Titrimetric test in which total, dissolved and hydrogen sulfide can be separated
Manufacturer:  LaMotte™ 4630/DEL
Catalog No. 22-307-862
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Dropper Pipet - Test uses a pipet to deliver standard size drops; the number of drops used to obtain a color change is multiplied by a given factor to produce the test result
  • Total, Dissolved and Hydrogen Sulfide - Test uses Pomeroy blue method for analysis
Specifications
Specifications
1ppm
Total Dissolved and Hydrogen Sulfide Kit
70.0
Measures for total, dissolved and hydrogen sulfide in drinking water, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Pomeroy Methylene Blue