Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
LaMotte™ Total Hardness Test Kit
Uses EDTA titration
Manufacturer:  LaMotte™ 4482LT02/DEL
Catalog No. 22-307-861
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Dropper Bottles - Bottle tips deliver a consistent standard drop size to add titrant to the sample; the number of drops used to complete the reaction is multiplied by a given equivalence factor to determine the concentration.
  • Hardness - Test uses EDTA titration with a red to blue endpoint; with inhibitors to eliminate metal interferences. Liquid buffer and tablet indicator. Results as CaCO3.
Specifications
Specifications
10ppm
Total Hardness Test Kit
50.0
Measures for total hardness in food and beverage, industrial water boiling and cooling, laundry, sanitation, water and wastewater
EDTA Titration