Learn More
LAMP-1/CD107a Mouse anti-Human, Mouse, Primate, Alexa Fluor™ 350, Clone: H4A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225183AF350
Catalog No. NP22183A30
Add to cart
Description
Description
LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human,Mouse,Primate samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
LAMP-1/CD107a
H4A3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody H4A3 recognizes CD107a, an approximately 100-120 kDa glycoprotein expressed mainly on lysosomal, but also on the plasma membrane.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
LAMP1
Human PBMC
0.1 mL
Primary
3916
Human, Mouse, Primate