Learn More
Abnova™ LAMP3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00027074T01
Catalog No. 89-950-366
Add to cart
Specifications
Specifications
293T
BC032940.2
pCMV-LAMP3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LAMP3 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 44.3kDa
Transient overexpression cell lysate was tested with anti-LAMP3 antibody (Mfr. No. H00027074-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human