Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANCL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LANCL1
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence -1:100-1:500, Immunoprecipitation -1-5-ul/mg
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121487
View Documents
Novus Biologicals
NBP31138950UL
50 μg
Each for $138.37
Add to cart
 
NB121486
View Documents
Novus Biologicals
NBP311389200UL
200 μg
Each for $320.71
Add to cart
 
Description

Description

LANCL1 Polyclonal antibody specifically detects LANCL1 in Human samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
Specifications

Specifications

LANCL1
Western Blot, Immunofluorescence, Immunoprecipitation
Purified
RUO
PBS, pH7.0
10314
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence -1:100-1:500, Immunoprecipitation -1-5-ul/mg
Unconjugated
Rabbit
Human
40 kDa erythrocyte membrane protein, LanC (bacterial lantibiotic synthetase component C)-like 1, LanC (bacterial lantibiotic synthetase component), LanC lantibiotic synthetase component C-like 1 (bacterial), lanC-like protein 1, p40GPR69AG protein-coupled receptor 69A
E. coli-derived Human LANCL1 fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit